Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Isoginkgetin

Catalog No. 648310
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-831-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 64-831-0 Supplier Tocris Bioscience™ Supplier No. 6483/10
This item is not returnable. View return policy

Pre-mRNA splicing inhibitor; cell permeable

Isoginkgetin is a pre-mRNA splicing inhibitor (IC50 ∽ 30 μM); inhibits major and minor spliceosomes. Suppresses transcriptional elongation. Blocks spliceosome-mediated splicing process at the pre-spliceosome/A complex stage. Does not inhibit CDK9. Inhibits tumor cell invasion. Active in vivo. Cell permeable

Specifications

Chemical Name or Material 8-[5-(5,7-Dihydroxy-4-oxo-4H-1-benzopyran-2-yl)-2-methoxyphenyl]-5,7-dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one
CAS 548-19-6
Quantity 10 mg
Molecular Formula C32H22O10
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.