accessibility menu, dialog, popup

UserName

Viewing profile as user:
FONNOHN

CAS RN 548-73-2

IUPAC Name: 1-{1-[4-(4-fluorophenyl)-4-oxobutyl]-1,2,3,6-tetrahydropyridin-4-yl}-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: droperidol inopsin droperidolum dehidrobenzperidol kern brand of droperidol droperidol (substance) droleptan properidol dridol innovar-vet
Molecular Weight (g/mol): 379.44
Molecular Formula: C22H22FN3O2
InChi Key: RMEDXOLNCUSCGS-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)CCCN1CCC(=CC1)N1C(=O)NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results