accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3IOOH

CAS RN 54811-38-0

IUPAC Name: 5-iodo-2-methylbenzoic acid
Synonyms: 5-iodo-2-methylbenzoic acid
Molecular Weight (g/mol): 262.05
Molecular Formula: C8H7IO2
InChi Key: WUBHOZQZSHGUFI-UHFFFAOYSA-N
SMILES: CC1=C(C=C(I)C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results