accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54815-13-3

IUPAC Name: 1,1-diethoxynonane
Synonyms: 1,1-diethoxynonane
Molecular Weight (g/mol): 216.37
Molecular Formula: C13H28O2
InChi Key: RJMSGTVQHHFVLK-UHFFFAOYSA-N
SMILES: CCCCCCCCC(OCC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results