Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonanal Diethyl Acetal 98.0+%, TCI America™
SDP

Catalog No. N09565ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09565ML 5 mL
N095625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09565ML Supplier TCI America Supplier No. N09565ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54815-13-3
Molecular Formula C13H28O2
Molecular Weight (g/mol) 216.37
MDL Number MFCD00672806
InChI Key RJMSGTVQHHFVLK-UHFFFAOYSA-N
Synonym 1,1-Diethoxynonane, Nonyl Aldehyde Diethyl Acetal
PubChem CID 108625
IUPAC Name 1,1-diethoxynonane
SMILES CCCCCCCCC(OCC)OCC

Specifications

Color Colorless
Boiling Point 66°C
Quantity 5 mL
Formula Weight 216.37
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonanal Diethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.