accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NI

CAS RN 54899-89-7

IUPAC Name: 1-(3-methylbutyl)quinolin-1-ium iodide
Synonyms: 1-(3-methylbutyl)quinolin-1-ium iodide
Molecular Weight (g/mol): 327.21
Molecular Formula: C14H18IN
InChi Key: JAHQMMYIQANKBZ-UHFFFAOYSA-M
SMILES: [I-].CC(C)CC[N+]1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results