accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNCH3H3COOHNCl

CAS RN 55-45-8

IUPAC Name: 4-(trimethylazaniumyl)but-2-yn-1-yl N-(3-chlorophenyl)carbamate chloride
Synonyms: 4-(trimethylammonio)but-2-yn-1-yl N-(3-chlorophenyl)carbamate chloride
Molecular Weight (g/mol): 317.21
Molecular Formula: C14H18Cl2N2O2
InChi Key: CXFZFEJJLNLOTA-UHFFFAOYSA-N
SMILES: [Cl-].C[N+](C)(C)CC#CCOC(=O)NC1=CC=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results