accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOOSCH3OO

CAS RN 55-98-1

IUPAC Name: 4-(methanesulfonyloxy)butyl methanesulfonate
Synonyms: busulfan busulfex wellcome, busulfan leucosulfan glaxo wellcome brand of busulfan myeleukon mielucin citosulfan BSF BUS
Molecular Weight (g/mol): 246.29
Molecular Formula: C6H14O6S2
InChi Key: COVZYZSDYWQREU-UHFFFAOYSA-N
SMILES: CS(=O)(=O)OCCCCOS(C)(=O)=O
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)