Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Butanediol Dimethanesulfonate 97.0+%, TCI America™
SDP

Catalog No. B102225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B102225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B102225G Supplier TCI America Supplier No. B102225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 55-98-1
Molecular Formula C6H14O6S2
Molecular Weight (g/mol) 246.292
MDL Number MFCD00007562
InChI Key COVZYZSDYWQREU-UHFFFAOYSA-N
Synonym busulfan, myleran, busulphan, sulphabutin, leucosulfan, myelosan, busulfex, citosulfan, mielucin, misulban
PubChem CID 2478
ChEBI CHEBI:28901
IUPAC Name 4-methylsulfonyloxybutyl methanesulfonate
SMILES CS(=O)(=O)OCCCCOS(=O)(=O)C

Specifications

Melting Point 118°C
Quantity 25 g
UN Number 2811
Formula Weight 246.29
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Butanediol Dimethanesulfonate
EINECSNumber (2)-1585
RTECSNumber EK1750000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.