accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClNHOCH3(E/Z)(E/Z)

CAS RN 55224-94-7

IUPAC Name: N-[4-(4-chlorophenyl)-3-methylbut-3-en-2-ylidene]hydroxylamine
Synonyms: N-[4-(4-chlorophenyl)-3-methylbut-3-en-2-ylidene]hydroxylamine
Molecular Weight (g/mol): 209.67
Molecular Formula: C11H12ClNO
InChi Key: MHTJEUOFLVQMCL-UHFFFAOYSA-N
SMILES: CC(=CC1=CC=C(Cl)C=C1)C(C)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results