accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5525-95-1

IUPAC Name: (2,3,4,5,6-pentafluorophenyl)diphenylphosphane
Synonyms: (2,3,4,5,6-pentafluorophenyl)diphenylphosphane
Molecular Weight (g/mol): 352.24
Molecular Formula: C18H10F5P
InChi Key: KUTXTUCJQJPJBH-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(P(C2=CC=CC=C2)C2=CC=CC=C2)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results