accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CSOO(R)(S)CH3CH2(S)OH(R)CH3(R)H3CH3C(S)OOHOOHO(E)

CAS RN 55297-96-6

IUPAC Name: (2E)-but-2-enedioic acid; (2R,3S,4S,6R,8S,14R)-4-ethenyl-3-hydroxy-2,4,7,14-tetramethyl-9-oxotricyclo[5.4.3.0¹,⁸]tetradecan-6-yl 2-{[2-(diethylamino)ethyl]sulfanyl}acetate
Synonyms: (2R,3S,4S,6R,8S,14R)-4-ethenyl-3-hydroxy-2,4,7,14-tetramethyl-9-oxotricyclo[5.4.3.0¹,⁸]tetradecan-6-yl 2-{[2-(diethylamino)ethyl]sulfanyl}acetate; fumaric acid
Molecular Weight (g/mol): 609.82
Molecular Formula: C32H51NO8S
InChi Key: YXQXDXAHCSEVSD-LKNSULBJSA-N
SMILES: OC(=O)\C=C\C(O)=O.CCN(CC)CCSCC(=O)O[C@@H]1C[C@@](C)(C=C)[C@@H](O)[C@H](C)C23CCC(=O)[C@@H]2C1(C)[C@H](C)CC3
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)