Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tiamulin Fumarate, G-Biosciences
SCI_ED

Catalog No. S139812
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139812 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139812 Supplier G-Biosciences Supplier No. RC1118
Add to Cart
Add to Cart

Tiamulin fumarate is a pleuromutilin antibiotic targeting gram-positive bacteria and several Mycoplasma species

Pleuromutilin antibiotics target the 50s ribosomal subunit, inhibiting protein synthesis in bacteria, by binding peptidyl transferase. This blocks formation of peptide bonds.

Chemical Identifiers

CAS 55297-96-6
Molecular Weight (g/mol) 609.81 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 1 g
Percent Purity >95%
Grade Molecular Biology
Chemical Name or Material Tiamulin Fumarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.