CAS RN 553-24-2
Thermo Scientific Chemicals Neutral Red
CAS: 553-24-2 Molecular Formula: C15H17ClN4 Molecular Weight (g/mol): 288.779 MDL Number: MFCD00012651 InChI Key: PGSADBUBUOPOJS-UHFFFAOYSA-N Synonym: Basic Red 5; C.I. 50040 PubChem CID: 11105 ChEBI: CHEBI:86370 IUPAC Name: 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride SMILES: CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
Thermo Scientific Chemicals Neutral Red, ACS
CAS: 553-24-2 Molecular Formula: C15H17ClN4 Molecular Weight (g/mol): 288.779 MDL Number: MFCD00012651 InChI Key: PGSADBUBUOPOJS-UHFFFAOYSA-N Synonym: Basic Red 5; C.I. 50040 PubChem CID: 11105 ChEBI: CHEBI:86370 IUPAC Name: 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride SMILES: CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
Thermo Scientific Chemicals Neutral Red, pure, high purity biological stain
CAS: 553-24-2 Molecular Formula: C15H17ClN4 Molecular Weight (g/mol): 288.779 MDL Number: MFCD00012651 InChI Key: PGSADBUBUOPOJS-UHFFFAOYSA-N Synonym: Basic Red 5,C.I. 50040,3-Amino-7-dimethylamino-2-methylphenazine hydrochloride PubChem CID: 11105 ChEBI: CHEBI:86370 IUPAC Name: 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride SMILES: CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
Basic Red 5, TCI America™
CAS: 553-24-2 Molecular Formula: C15H17ClN4 Molecular Weight (g/mol): 288.779 MDL Number: MFCD00012651 InChI Key: PGSADBUBUOPOJS-UHFFFAOYSA-N Synonym: Neutral Red, Toluylene Red PubChem CID: 11105 ChEBI: CHEBI:86370 IUPAC Name: 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride SMILES: CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
Neutral Red, Reagent, ACS, Spectrum™ Chemical
Thermo Scientific Chemicals Neutral Red, pure, certified
CAS: 553-24-2 Molecular Formula: C15H17ClN4 Molecular Weight (g/mol): 288.779 MDL Number: MFCD00012651 InChI Key: PGSADBUBUOPOJS-UHFFFAOYSA-N Synonym: Basic Red 5,C.I. 50040,3-Amino-7-dimethylamino-2-methylphenazine hydrochloride PubChem CID: 11105 ChEBI: CHEBI:86370 IUPAC Name: 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride SMILES: CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
Neutral Red TS, 0.1% (w/v) in 50% Alcohol, Ricca Chemical
CAS: 553-24-2 Molecular Formula: C15H17ClN4 Molecular Weight (g/mol): Mixture InChI Key: PGSADBUBUOPOJS-UHFFFAOYSA-N PubChem CID: 11105 ChEBI: CHEBI:86370
Neutral Red, 1% AQ, Vion Biosciences
CAS: 553-24-2 Molecular Formula: C15H17ClN4 Molecular Weight (g/mol): 288.78 MDL Number: MFCD00012651 InChI Key: PGSADBUBUOPOJS-UHFFFAOYSA-N Synonym: Neutral Red, Water Solution PubChem CID: Data not available. ChEBI: CHEBI:86370 IUPAC Name: hydrogen N2,N2,7-trimethylphenazine-2,8-diamine chloride SMILES: [H+].[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=NC2=C1
Neutral Red (Certified Biological Stain), Fisher Chemical
CAS: 553-24-2 Molecular Formula: C15H17ClN4 Molecular Weight (g/mol): 288.779 MDL Number: MFCD00012651 InChI Key: PGSADBUBUOPOJS-UHFFFAOYSA-N Synonym: Basic Red 5,Toluylene Red PubChem CID: 11105 ChEBI: CHEBI:86370 IUPAC Name: 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride SMILES: CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
MilliporeSigma™ Neutral Red Stain
Stains bacteriological sample material of human origin.