accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OCl

CAS RN 55312-97-5

IUPAC Name: 2-(2,5-dimethylphenyl)acetyl chloride
Synonyms: 2-(2,5-dimethylphenyl)acetyl chloride
Molecular Weight (g/mol): 182.65
Molecular Formula: C10H11ClO
InChi Key: PZRANTBLOIKHJY-UHFFFAOYSA-N
SMILES: CC1=CC(CC(Cl)=O)=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results