accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(R)N(S)O(S)(R)NNOHNH2HOHO

CAS RN 5536-17-4

IUPAC Name: 2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: adenosine
Molecular Weight (g/mol): 267.25
Molecular Formula: C10H13N5O4
InChi Key: OIRDTQYFTABQOQ-UHFFFAOYNA-N
SMILES: NC1=C2N=CN(C3OC(CO)C(O)C3O)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results