accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONOCl

CAS RN 55435-71-7

IUPAC Name: 4-(2-chloro-4-nitrophenyl)morpholine
Synonyms: 4-(2-chloro-4-nitrophenyl)morpholine
Molecular Weight (g/mol): 242.66
Molecular Formula: C10H11ClN2O3
InChi Key: ZPKGWYXJULKUEH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(N2CCOCC2)C(Cl)=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)