accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3SiH3CCH3FFFFF

CAS RN 55485-74-0

IUPAC Name: [dimethyl(2,3,4,5,6-pentafluorophenyl)silyl]diethylamine
Synonyms: [dimethyl(2,3,4,5,6-pentafluorophenyl)silyl]diethylamine
Molecular Weight (g/mol): 297.34
Molecular Formula: C12H16F5NSi
InChi Key: ZBVPABKIRWOTNJ-UHFFFAOYSA-N
SMILES: CCN(CC)[Si](C)(C)C1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results