accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFBFFFFSSSSSSSSSSSS

CAS RN 55492-86-9

IUPAC Name: 2-(1λ⁴,3-dithiol-1-ylium-2-yl)-1λ⁴,3-dithiol-1-ylium; bis(2-(2H-1,3-dithiol-2-ylidene)-2H-1,3-dithiole); bis(tetrafluoroboranuide)
Synonyms: 2-(1λ⁴,3-dithiol-1-ylium-2-yl)-1λ⁴,3-dithiol-1-ylium bis(tetrathiafulvalene) ditetrafluoroborate
Molecular Weight (g/mol): 786.62
Molecular Formula: C18H12B2F8S12
InChi Key: IJZRAQUJMWAFMU-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.F[B-](F)(F)F.S1C=CSC1=C1SC=CS1.S1C=CSC1=C1SC=CS1.S1C=C[S+]=C1C1=[S+]C=CS1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results