accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 555-30-6

IUPAC Name: (2S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoic acid
Synonyms: methyldopa becanta methyldopum clonmel brand of methyldopa nu-pharm brand of methyldopa methyl-L-dopa α medopa methyldopa orion brand nu-medopa hypolag
Molecular Weight (g/mol): 211.22
Molecular Formula: C10H13NO4
InChi Key: CJCSPKMFHVPWAR-JTQLQIEISA-N
SMILES: C[C@](N)(CC1=CC=C(O)C(O)=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results