CAS RN 55661-38-6
IUPAC Name:
hydrogen 1-[(4-amino-2-methylpyrimidin-5-yl)methyl]-3-(2-chloroethyl)-3-nitrosourea chloride
Synonyms:
hydrogen nimustine chloride
Molecular Weight (g/mol):
309.15
Molecular Formula:
C9H14Cl2N6O2
InChi Key:
KPMKNHGAPDCYLP-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CC1=NC=C(CNC(=O)N(CCCl)N=O)C(N)=N1