Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nimustine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. N08211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08211G Supplier TCI America Supplier No. N08211G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 55661-38-6
Molecular Formula C9H14Cl2N6O2
Molecular Weight (g/mol) 309.15
MDL Number MFCD01676942
InChI Key KPMKNHGAPDCYLP-UHFFFAOYSA-N
Synonym ACNU, 1-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-3-(2-chloroethyl)-3-nitrosourea Hydrochloride
PubChem CID 91657
ChEBI CHEBI:7576
IUPAC Name hydrogen 1-[(4-amino-2-methylpyrimidin-5-yl)methyl]-3-(2-chloroethyl)-3-nitrosourea chloride
SMILES [H+].[Cl-].CC1=NC=C(CNC(=O)N(CCCl)N=O)C(N)=N1

Specifications

Melting Point 186°C
Color Yellow
Quantity 1 g
UN Number 2811
Formula Weight 309.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nimustine Hydrochloride
RTECSNumber YR8450000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.