accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSSHN

CAS RN 55690-60-3

IUPAC Name: 5-methoxy-2,3-dihydro-1,3-benzothiazole-2-thione
Synonyms: 5-methoxy-3H-1,3-benzothiazole-2-thione
Molecular Weight (g/mol): 197.27
Molecular Formula: C8H7NOS2
InChi Key: JDPITNFDYXOKRM-UHFFFAOYSA-N
SMILES: COC1=CC=C2SC(=S)NC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results