Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-5-methoxybenzothiazole 98.0+%, TCI America™
SDP

Catalog No. M12775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12775G Supplier TCI America Supplier No. M12775G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55690-60-3
Molecular Formula C8H7NOS2
Molecular Weight (g/mol) 197.27
MDL Number MFCD00185941
InChI Key JDPITNFDYXOKRM-UHFFFAOYSA-N
Synonym 5-methoxybenzo d thiazole-2-thiol, 2-mercapto-5-methoxybenzothiazole, 5-methoxybenzothiazole-2-thiol, 5-methoxy-1,3-benzothiazole-2-thiol, 5-methoxybenzothiazole-2 3h-thione, 5-methoxy-2-benzothiazolethiol, 5-methoxy-2-mercaptobenzothiazole, 2-mercapto-5-methoxy-benzothiazole, acmc-209loj, 2-mercapto5-methoxybenzothiazole
PubChem CID 2830679
IUPAC Name 5-methoxy-2,3-dihydro-1,3-benzothiazole-2-thione
SMILES COC1=CC=C2SC(=S)NC2=C1

Specifications

Melting Point 198°C
Color Yellow
Quantity 5 g
Formula Weight 197.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercapto-5-methoxybenzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.