accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3C(R)(R)(S)CH3(R)O(S)CH3(S)HO(S)CH3O(R)CH3(S)O(R)O(S)(S)CH3O(R)(R)OHCH3(S)CH3O(R)OH(R)CH3(S)H3COHO

CAS RN 55721-31-8

IUPAC Name: sodium (2R)-2-[(2R,5S,6R)-6-[(2S,3S,4S,6R)-6-[(2S,5S,7R,9S,10S,12R,15R)-2-[(2R,5R,6S)-5-ethyl-5-hydroxy-6-methyloxan-2-yl]-15-hydroxy-2,10,12-trimethyl-1,6,8-trioxadispiro[4.1.5⁷.3⁵]pentadec-13-en-9-yl]-3-hydroxy-4-methyl-5-oxooctan-2-yl]-5-methyloxan-2-yl]butanoic acid
Synonyms: salinomycin
Molecular Weight (g/mol): 774
Molecular Formula: C42H70NaO11
InChi Key: YPZYGIQXBGHDBH-UZHRAPRISA-N
SMILES: [Na+].CC[C@H]([C@H]1CC[C@H](C)[C@@H](O1)[C@@H](C)[C@H](O)[C@H](C)C(=O)[C@H](CC)[C@H]1O[C@@]2(O[C@@]3(CC[C@](C)(O3)[C@H]3CC[C@](O)(CC)[C@H](C)O3)[C@H](O)C=C2)[C@H](C)C[C@@H]1C)C(O)=O
Grade 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Color 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)