accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 55726-22-2

IUPAC Name: icosanoyl icosanoate
Synonyms: icosanoyl icosanoate
Molecular Weight (g/mol): 607.06
Molecular Formula: C40H78O3
InChi Key: AUONNNVJUCSETH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results