accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5597-27-3

IUPAC Name: 3-methylidenebicyclo[2.2.1]heptan-2-one
Synonyms: 3-methylidenebicyclo[2.2.1]heptan-2-one
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: FNOOZJAPZFHNCW-UHFFFAOYNA-N
SMILES: C=C1C2CCC(C2)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results