accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaHOOClOO

CAS RN 56047-23-5

IUPAC Name: sodium 2-carboxy-5-chlorobenzoate
Synonyms: sodium 2-carboxy-5-chlorobenzoate
Molecular Weight (g/mol): 222.56
Molecular Formula: C8H4ClNaO4
InChi Key: MWZMHSGEQHVWED-UHFFFAOYSA-M
SMILES: [Na+].OC(=O)C1=CC=C(Cl)C=C1C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results