accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56051-06-0

CH3CH3SeCH3SeH3C

CAS RN 56051-06-0

IUPAC Name: 2,2-bis(methylselanyl)propane
Synonyms: 2,2-bis(methylselanyl)propane
Molecular Weight (g/mol): 230.09
Molecular Formula: C5H12Se2
InChi Key: KPGCXISRRPRULE-UHFFFAOYSA-N
SMILES: C[Se]C(C)(C)[Se]C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results