accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56106-37-7

H3COOCH3

CAS RN 56106-37-7

IUPAC Name: 1,3-dipropoxybenzene
Synonyms: 1,3-dipropoxybenzene
Molecular Weight (g/mol): 194.27
Molecular Formula: C12H18O2
InChi Key: UTFRNSPYRPYKDV-UHFFFAOYSA-N
SMILES: CCCOC1=CC(OCCC)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results