accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOOOOOOH

CAS RN 5617-32-3

IUPAC Name: 3,6,9,12,15,18-hexaoxaicosane-1,20-diol
Synonyms: heptaethylene glycol, peg330 peg330 heptaethylene glycol
Molecular Weight (g/mol): 326.39
Molecular Formula: C14H30O8
InChi Key: XPJRQAIZZQMSCM-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCCOCCOCCOCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results