accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56175-44-1

IUPAC Name: 1-methoxy-2,3-dihydro-1H-inden-2-ol
Synonyms: 1-methoxy-2,3-dihydro-1H-inden-2-ol
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: ZHWPZRVYPHGXSF-UHFFFAOYNA-N
SMILES: COC1C(O)CC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results