accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClClClCl

CAS RN 56207-45-5

IUPAC Name: 2,2,6,6-tetrachlorocyclohexan-1-ol
Synonyms: 2,2,6,6-tetrachlorocyclohexan-1-ol
Molecular Weight (g/mol): 237.93
Molecular Formula: C6H8Cl4O
InChi Key: RORBTKDJFQCFMD-UHFFFAOYSA-N
SMILES: OC1C(Cl)(Cl)CCCC1(Cl)Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results