accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3(Z)

CAS RN 56219-06-8

IUPAC Name: methyl (9Z)-tetradec-9-enoate
Synonyms: methyl (9Z)-tetradec-9-enoate
Molecular Weight (g/mol): 240.39
Molecular Formula: C15H28O2
InChi Key: RWIPSJUSVXDVPB-SREVYHEPSA-N
SMILES: CCCC\C=C/CCCCCCCC(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results