Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myristoleic Acid Methyl Ester, MP Biomedicals™

Catalog No. MP215576190 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215576190 500 mg
MP215576150 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215576190 Supplier MP Biomedicals, Inc Supplier No. 0215576190
Add to Cart
Add to Cart

Induces apoptosis and necrosis in human prostate cancer LNCaP cells. Myristoleic Acid Methyl Ester, MP Biomedicals is a cytotoxic component from the fruit extract of S. repens. It is a more hydrophobic form of the free acid.

Myristoleic acid induces apoptosis and necrosis in human prostate cancer LNCaP cells at a rate of 8.8% and 8.1%, respectively. It has a minimal inhibitory concentration (MIC) of 9μM in vivo.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56219-06-8
Molecular Formula C15H28O2
Molecular Weight (g/mol) 240.387
InChI Key RWIPSJUSVXDVPB-SREVYHEPSA-N
Synonym methyl myristoleate, myristoleic acid methyl ester, methyl cis-9-tetradecenoate, unii-m2a58gy964, z-methyl tetradec-9-enoate, methyl z-tetradec-9-enoate, cis-9-tetradecenoic acid, methyl ester, methyl 9z-tetradec-9-enoate, formyl 9z-tetradecenoate, methyl 9z-tetradecenoate
PubChem CID 5352674
IUPAC Name methyl (Z)-tetradec-9-enoate
SMILES CCCCC=CCCCCCCCC(=O)OC

Specifications

Refractive Index n20/D 1.446 (Lit.)
Quantity 500 mg
Formula Weight 240.4
Percent Purity ≥99%
Physical Form Liquid
Recommended Storage −20°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.