Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myristoleic Acid Methyl Ester, 99%, MP Biomedicals™

Catalog No. ICN15576180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15576180 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15576180 Supplier MP Biomedicals, Inc Supplier No. 0215576180
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56219-06-8
Molecular Formula C15H28O2
Molecular Weight (g/mol) 240.387
InChI Key RWIPSJUSVXDVPB-SREVYHEPSA-N
Synonym methyl myristoleate, myristoleic acid methyl ester, methyl cis-9-tetradecenoate, unii-m2a58gy964, z-methyl tetradec-9-enoate, methyl z-tetradec-9-enoate, cis-9-tetradecenoic acid, methyl ester, methyl 9z-tetradec-9-enoate, formyl 9z-tetradecenoate, methyl 9z-tetradecenoate
PubChem CID 5352674
IUPAC Name methyl (Z)-tetradec-9-enoate
SMILES CCCCC=CCCCCCCCC(=O)OC

Specifications

Assay Percent Range 99%
Quantity 100 mg
Formula Weight 240.4
Physical Form Liquid
Chemical Name or Material Myristoleic acid methyl ester
Recommended Storage Store at 0°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.