accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56452-52-9

IUPAC Name: bis(2-amino-3-(1H-indol-3-yl)-2-methylpropanoic acid) hydrate
Synonyms: bis(2-amino-3-(1H-indol-3-yl)-2-methylpropanoic acid) hydrate
Molecular Weight (g/mol): 454.53
Molecular Formula: C24H30N4O5
InChi Key: BZCFHAYHUKOGKE-IUWNWRFPSA-N
SMILES: O.CC(N)(CC1=CNC2=CC=CC=C12)C(O)=O.CC(N)(CC1=CNC2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results