accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5666-17-1

IUPAC Name: diethyl(prop-2-en-1-yl)amine
Synonyms: diethyl(prop-2-en-1-yl)amine
Molecular Weight (g/mol): 113.2
Molecular Formula: C7H15N
InChi Key: JWAJUTZQGZBKFS-UHFFFAOYSA-N
SMILES: CCN(CC)CC=C
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results