Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylallylamine 98.0+%, TCI America™
SDP

Catalog No. D118525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D118525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D118525ML Supplier TCI America Supplier No. D118525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5666-17-1
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.204
MDL Number MFCD00048537
InChI Key JWAJUTZQGZBKFS-UHFFFAOYSA-N
Synonym n,n-diethylallylamine, allyldiethylamine, n-allyldiethylamine, 2-propen-1-amine, n,n-diethyl, diethylallylamine, diethyl prop-2-en-1-yl amine, n,n-diethyl-2-propen-1-amine, allyldiethyl amine, allyl diethyl amine, allyl-diethyl-amine
PubChem CID 79748
IUPAC Name N,N-diethylprop-2-en-1-amine
SMILES CCN(CC)CC=C

Specifications

Color Colorless
Boiling Point 110°C
Quantity 25 mL
UN Number 2733
Formula Weight 113.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Diethylallylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.