Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyldiethylamine, MP Biomedicals

Catalog No. ICN202933 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202933 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202933 Supplier MP Biomedicals, Inc Supplier No. 0520293310

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5666-17-1
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.204
InChI Key JWAJUTZQGZBKFS-UHFFFAOYSA-N
Synonym n,n-diethylallylamine, allyldiethylamine, n-allyldiethylamine, 2-propen-1-amine, n,n-diethyl, diethylallylamine, diethyl prop-2-en-1-yl amine, n,n-diethyl-2-propen-1-amine, allyldiethyl amine, allyl diethyl amine, allyl-diethyl-amine
PubChem CID 79748
IUPAC Name N,N-diethylprop-2-en-1-amine
SMILES CCN(CC)CC=C

Specifications

Quantity 10 g
Formula Weight 113.2
Chemical Name or Material Allyldiethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.