accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNOBrFFF

CAS RN 56661-32-6

IUPAC Name: 4-bromo-N-[3-(trifluoromethyl)phenyl]benzamide
Synonyms: 4-bromo-N-[3-(trifluoromethyl)phenyl]benzamide
Molecular Weight (g/mol): 344.13
Molecular Formula: C14H9BrF3NO
InChi Key: URMINAWTHPPDQD-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(NC(=O)C2=CC=C(Br)C=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results