accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOOFFF

CAS RN 5672-89-9

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2,2,2-trifluoroacetate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2,2,2-trifluoroacetate
Molecular Weight (g/mol): 211.1
Molecular Formula: C6H4F3NO4
InChi Key: NIANZCQUVKPGSF-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)ON1C(=O)CCC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results