accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3H3C

CAS RN 5673-07-4

IUPAC Name: 1,3-dimethoxy-2-methylbenzene
Synonyms: 1,3-dimethoxy-2-methylbenzene
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: FPEUDBGJAVKAEE-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OC)=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results