accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5675-25-2

IUPAC Name: (4E)-octa-1,4-diene
Synonyms: (4E)-octa-1,4-diene
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: HYBLFDUGSBOMPI-BQYQJAHWSA-N
SMILES: CCC\C=C\CC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results