accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5676-48-2

OOHOOHOHOHH2OH2O

CAS RN 5676-48-2

IUPAC Name: tetrahydroxycyclohexa-2,5-diene-1,4-dione dihydrate
Synonyms: kelox dihydrate
Molecular Weight (g/mol): 208.12
Molecular Formula: C6H8O8
InChi Key: PTIMJXRLFTZAOV-UHFFFAOYSA-N
SMILES: O.O.OC1=C(O)C(=O)C(O)=C(O)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results