accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOOIOOIOII(?)

CAS RN 568-63-8

IUPAC Name: disodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Synonyms: erythrosine
Molecular Weight (g/mol): 879.86
Molecular Formula: C20H6I4Na2O5
InChi Key: IINNWAYUJNWZRM-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12
Quantity 
  • (1)
  • (1)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results