Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythrosin B Sodium Salt, MP Biomedicals™

Catalog No. MP219044905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219044905 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219044905 Supplier MP Biomedicals, Inc Supplier No. 0219044905
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as a contrast or counter stain to hematoxylin, crystal violet and other nuclear stains. Erythrosin B Sodium Salt, MP Biomedicals is a red, anionic xanthene dye for use with both plant and animal tissue. Erythosin B solution in 0.9% saline is made for

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 568-63-8
Molecular Formula C40H14I8Na2O10
Molecular Weight (g/mol) 1715.757
InChI Key DKRYETBOLGDIOK-UHFFFAOYSA-L
Synonym Acid Red 51, Solvent Red 140, 2',4',5',7'-Tetraiodofluorescein, FD&C Red No. 3, Erythrosin extra bluish
PubChem CID 131676154
IUPAC Name disodium;2-(3-hydroxy-2,4,5,7-tetraiodo-6-oxoxanthen-9-yl)benzoic acid;2-(2,4,5,7-tetraiodo-3-oxido-6-oxoxanthen-9-yl)benzoate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)O)I)I)I)C(=O)O.C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)[O-])I)I)I)C(=O)[O-].[Na+].[Na+]

Specifications

Solubility Information Soluble in water (20mg/mL) or ethanol (20mg/mL)
Quantity 5 g
Physical Form Powder
Formula Weight 879.9
Recommended Storage Store at Room Temperature (15-30°C), desiccate.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.