Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythrosin B, ∽93% by uv-vis, MP Biomedicals™

Catalog No. ICN19044925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19044925 25 g
ICN19044910 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19044925 Supplier MP Biomedicals, Inc Supplier No. 0219044925
Add to Cart
Add to Cart

Chemical Identifiers

CAS 568-63-8
Molecular Formula C40H14I8Na2O10
Molecular Weight (g/mol) 1715.757
MDL Number MFCD00144257
InChI Key DKRYETBOLGDIOK-UHFFFAOYSA-L
Synonym Acid Red 51, C.I. 45430
PubChem CID 131676154
IUPAC Name disodium;2-(3-hydroxy-2,4,5,7-tetraiodo-6-oxoxanthen-9-yl)benzoic acid;2-(2,4,5,7-tetraiodo-3-oxido-6-oxoxanthen-9-yl)benzoate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)O)I)I)I)C(=O)O.C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)[O-])I)I)I)C(=O)[O-].[Na+].[Na+]

Specifications

Assay Percent Range ∽ 93%
Formula Weight 879.9
Color Maroon
Quantity 25 g
Physical Form Powder
Chemical Name or Material Erythrosin B Sodium Salt
Recommended Storage Store at room temperature (15° to 30°C) and desiccated.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.