accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHHOOOOOOH

CAS RN 568-80-9

IUPAC Name: 6,8,9,10-tetrahydroxy-2H,3H-pyrano[3,2-c]isochromene-2,3-dione
Synonyms: 6,8,9,10-tetrahydroxypyrano[3,2-c]isochromene-2,3-dione
Molecular Weight (g/mol): 278.17
Molecular Formula: C12H6O8
InChi Key: JWDHCGIDCCHUKI-UHFFFAOYSA-N
SMILES: OC1=C2C=C(O)C(O)=C(O)C2=C2OC(=O)C(=O)C=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)