Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Galloflavin
SDP

Catalog No. 479510 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-951-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 47-951-0 Supplier R&D Systems™ Supplier No. 4795/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Lactate dehydrogenase inhibitor; impairs aerobic glycolysis

Inhibitor of human lactate dehydrogenase (LDH) (Ki values are 5.46 and 15.1 μM for LDH-A and LDH-B, respectively, in competition with pyruvate). Inhibits lactate production and decreases ATP synthesis in PLD/PRF/5 cells; thought to decrease cell viability via impairment of aerobic glycolysis.

Chemical Identifiers

CAS 568-80-9
Molecular Formula C12H6O8

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 316.26
Purity >95%
Molecular Weight (g/mol) 278.17
Chemical Name or Material Galloflavin
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.